Learn More
Biochain Institute Inc Paraffin Tissue Section Panel - Human Adult Normal Tissue, Multi-tissue (8) II: Major Organ, 5 slides/PK
SDP
Catalog No. 50182507
Encompass_Preferred
BioChain's human adult normal multiple tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different human normal tissues. Tissues mounted on positively charged glass slides are organized either by organs (major organ multiple tissue panels), or by integrated major systems (organ systems multiple tissue panels). Documentation of tissues' clinical histories is available upon request. Identical format total RNA Northern Blots are available, and cDNA from those tissues may also be available for PCR detection.
Catalog No. 50-182-507 Supplier Biochain Institute Inc Supplier No. T8234432
BioChain's human adult normal multiple tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different human normal tissues. Tissues mounted on positively charged glass slides are organized either by organs (major organ multiple tissue panels), or by integrated major systems (organ systems multiple tissue panels). Documentation of tissues' clinical histories is available upon request. Identical format total RNA Northern Blots are available, and cDNA from those tissues may also be available for PCR detection.