Learn More
Biochain Institute Inc Paraffin Tissue Section Panel - Rat Normal Tissue, Multi-tissue I, 5 slides/PK
SDP
Catalog No. 50182557
Encompass_Preferred
BioChain's rat normal multiple tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different Sprague Dawley or Wistar rat tissues. Tissues are mounted on positively charged glass slides. Total RNA Northern Blots are available in the identical format, and cDNA from those tissues may also be available for PCR detection.
Catalog No. 50-182-557 Supplier Biochain Institute Inc Supplier No. T8434448
BioChain's rat normal multiple tissue panels are designed for studying tissue specific cellular localization of genes or proteins of interest in different Sprague Dawley or Wistar rat tissues. Tissues are mounted on positively charged glass slides. Total RNA Northern Blots are available in the identical format, and cDNA from those tissues may also be available for PCR detection.