Learn More
Biochain Institute Inc Paraffin Tissue Section Panel - Human Tumor, Different Tumor Type, Different Donors, Panel II, 5 tissue spots, 5 slides/PK
SDP
Catalog No. 50182526
Encompass_Preferred
BioChain's FFPE multiple tissue panels with different types of tumors are designed for screening gene expression in multiple tumor tissues. Each tumor tissue is paired with a corresponding normal control tissue. A licensed pathologist issued a pathological diagnosis of each tumor based on examination of tissue sections. Documentation of tissues' clinical histories is available upon request. mRNA Northern blot and Western blots as well as total RNA Norterhn blot are available in identical formats.
Catalog No. 50-182-526 Supplier Biochain Institute Inc Supplier No. T8235418
BioChain's FFPE multiple tissue panels with different types of tumors are designed for screening gene expression in multiple tumor tissues. Each tumor tissue is paired with a corresponding normal control tissue. A licensed pathologist issued a pathological diagnosis of each tumor based on examination of tissue sections. Documentation of tissues' clinical histories is available upon request. mRNA Northern blot and Western blots as well as total RNA Norterhn blot are available in identical formats.