Learn More
Mopec™ Paraffin Wax Block Trimmer
SDP
Catalog No. 22445536 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Paraffin Trimmer
Catalog No. Product Type
22-445-536 Paraffin Trimmer
1 options
Catalog No. 22-445-536 Supplier Mopec™ Supplier No. BK001
Add to Cart
Add to Cart
This Paraffin Wax Block Trimmer is designed to easily remove excess paraffin from the edges and writing surfaces of embedding tissue cassettes without any damage to written text or barcodes.
  • Rapid heating system and extra large work surface allows multiple cassettes to be de-waxed simultaneously
  • Features digital temperature control with memory function for desired start-up temperature
  • Neutral slope accessible for both left and right-hand users
Specifications
Product Type Paraffin Trimmer
For Use With (Equipment) Embedding Tissue Cassettes
For Use With (Application) Removing excess paraffin
Dimensions (L x W x H) 8 x 6 x 7.25 in.