Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paraffin Wax, Refined Gran, Vion Biosciences
SCI_ED

Catalog No. S02488 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02488 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02488 Supplier Aldon Corporation Supplier No. VNPP0056500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This paraffin wax is an adaptable embedding agent that provides superior preservation of morphology in histological specimens.

  • This paraffin wax is an adaptable embedding agent that provides superior preservation of morphology in histological specimens.

Specifications

Boiling Point Data not available.
Color White
Packaging Type HDPE Bottle
Melting Point 48.8°C to 71°C
Product Type Embedding Media
Quantity 500 g
For Use With (Application) Tissue Embedding Media
Material Paraffin Wax
Form Solid
Format Granules
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.