Learn More
Medicus Health Digital Paraffin Wax Trimmer
SDP
Catalog No. NC2173407
Encompass
∙Digital Wax Trimmer safely cleans the side of blocks so they will stay secure in the chuck ∙Features a digital control panel to monitor changes in heat level and to adjust temperature ∙ Designed to allow for trimming of multiple of multiple cassettes simultaneously to save operator time ∙ Collection tray stays in place by magnetic contact ∙ Our convenient PE disposable tray liner allows easy disposal of the runoff wax and grooved surface for proper wax drainage ∙ Temperature range: Room temperature - 120° C ∙ Power requirements: 150W ∙Includes 5 disposable drip tray liners ∙ Dimensions: 11.25"W x 5"D x 8"
This item is not returnable. View return policy
Catalog No. NC2173407 Supplier Medicus Health Supplier No. 5314M2
∙Digital Wax Trimmer safely cleans the side of blocks so they will stay secure in the chuck ∙Features a digital control panel to monitor changes in heat level and to adjust temperature ∙ Designed to allow for trimming of multiple of multiple cassettes simultaneously to save operator time ∙ Collection tray stays in place by magnetic contact ∙ Our convenient PE disposable tray liner allows easy disposal of the runoff wax and grooved surface for proper wax drainage ∙ Temperature range: Room temperature - 120° C ∙ Power requirements: 150W ∙Includes 5 disposable drip tray liners ∙ Dimensions: 11.25"W x 5"D x 8"