Learn More
Grainger Laboratory Film: Non-Sterile, Polyolefins and Paraffin Waxes, 250 ft Lg, 2 in Wd
Catalog No. NC1986430
Encompass_Preferred
Flexible and self-sealing material allows it to cling around irregular shapes and surfaces. Good resistance against acids, non-toxic and free from plasticizers.
Catalog No. NC1986430 Supplier Grainger Supplier No. 15G075
Flexible and self-sealing material allows it to cling around irregular shapes and surfaces. Good resistance against acids, non-toxic and free from plasticizers.