Learn More
Chemglass Life Sciences Chemglass Parafilm M sealing film, 2 in x 250 ft roll, 1 roll
SDP
Catalog No. NC2153658
Encompass_Preferred
Parafilm M sealing film provides a flexible, moisture-proof, self-adhering barrier for sealing laboratory glassware and containers. Supplied as a 2 in x 250 ft roll, it conforms to irregular shapes to reduce evaporation and contamination in sample handling applications.
  • Flexible, semi-transparent thermoplastic film that molds to surfaces.
  • Roll dimensions: 2 in x 250 ft; one roll per pack.
  • Stretches more than 200% for secure seals around irregular shapes.
  • Resistant to many acids, alkalis, and some organic solvents.
  • Not recommended for use with acetone, diethyl ether, or tetrachloride.
This item is not returnable. View return policy
Catalog No. NC2153658 Supplier Chemglass Life Sciences Supplier No. CG1398P050
Parafilm M sealing film provides a flexible, moisture-proof, self-adhering barrier for sealing laboratory glassware and containers. Supplied as a 2 in x 250 ft roll, it conforms to irregular shapes to reduce evaporation and contamination in sample handling applications.
  • Flexible, semi-transparent thermoplastic film that molds to surfaces.
  • Roll dimensions: 2 in x 250 ft; one roll per pack.
  • Stretches more than 200% for secure seals around irregular shapes.
  • Resistant to many acids, alkalis, and some organic solvents.
  • Not recommended for use with acetone, diethyl ether, or tetrachloride.