Learn More
Sigma Aldrich Fine Chemicals Biosciences PARAFILM M, Roll Size 4in x 125 ft.
Catalog No. NC1345631
Encompass
PARAFILM stretches up to 200% and clings even around irregular shapes and surfaces. The sealing film is free of plasticizers and consists primarily of polyolefins and paraffin waxes. PARAFILM M is resistant up to 48 hours against many polar substances, eg saline solutions, inorganic acids and alkaline solutions.
Catalog No. NC1345631 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P77931EA
PARAFILM stretches up to 200% and clings even around irregular shapes and surfaces. The sealing film is free of plasticizers and consists primarily of polyolefins and paraffin waxes. PARAFILM M is resistant up to 48 hours against many polar substances, eg saline solutions, inorganic acids and alkaline solutions.