Learn More
Polysciences, Inc. Parafilm, 4 in. x 250 ft./ROL, Used to seal laboratory vessels with a disposable, translucent film. Also used to stain E.M. grids
SDP
Catalog No. NC9834319
Encompass
Parafilm, 4 in. x 250 ft./ROL, Used to seal laboratory vessels with a disposable, translucent film. Also used to stain E.M. grids
Catalog No. NC9834319 Supplier Polysciences, Inc. Supplier No. 3989A1
Parafilm, 4 in. x 250 ft./ROL, Used to seal laboratory vessels with a disposable, translucent film. Also used to stain E.M. grids