Learn More
Chemglass Life Sciences Parafilm M Laboratory Sealing Film, 4 in x 125 ft Roll
SDP
Catalog No. NC2153659
Encompass_Preferred
Parafilm M is a flexible thermoplastic laboratory sealing film supplied as a 4 in x 125 ft roll. It stretches and conforms to irregular surfaces to form a temporary, watertight seal for tubes, flasks, beakers, and culture dishes, helping prevent evaporation and contamination during routine lab procedures.
  • Flexible thermoplastic film conforms to irregular surfaces.
  • Provides a moisture- and contamination-resistant seal to reduce evaporation.
  • Chemically resistant for short-term exposure to many acids, bases, and solvents.
  • Non-toxic, latex-free, and odor-free for general lab use.
  • Roll format provides long length per unit for frequent use.
This item is not returnable. View return policy
Catalog No. NC2153659 Supplier Chemglass Life Sciences Supplier No. CG1398P100
Parafilm M is a flexible thermoplastic laboratory sealing film supplied as a 4 in x 125 ft roll. It stretches and conforms to irregular surfaces to form a temporary, watertight seal for tubes, flasks, beakers, and culture dishes, helping prevent evaporation and contamination during routine lab procedures.
  • Flexible thermoplastic film conforms to irregular surfaces.
  • Provides a moisture- and contamination-resistant seal to reduce evaporation.
  • Chemically resistant for short-term exposure to many acids, bases, and solvents.
  • Non-toxic, latex-free, and odor-free for general lab use.
  • Roll format provides long length per unit for frequent use.