Learn More
Biomiga Inc. PARAFILM 4X125
Catalog No. NC9952494
Encompass_Preferred
It is used to further seal a lidded container against moisture and air contamination for long term storage
Catalog No. NC9952494 Supplier Biomiga Inc. Supplier No. BBPF002
It is used to further seal a lidded container against moisture and air contamination for long term storage