Learn More
Fisherbrand™ Parafilm™ Dispenser, ABS
Catalog No. 03448407 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Green
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
03-448-407 Blue
03-448-408 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-448-407
Add to Cart
Add to Cart
Store, dispense, and cut with ease.
  • The Parafilm™ dispenser has a unique design for maximum safety, storage, dispensing, and cutting of Parafilm™, tape and labels
  • ABS plastic dispenser designed with integrated safety razor that slides smoothly creating a clean cut
  • Supplied with two safety blades, additional replacement blades are available for purchase
  • Dispenser holds one or two 50 mm (2-in.) rolls or one 100 mm (4-in.) roll
Specifications
Product Type Parafilm Dispenser
Color Blue
Holds Parafilm
Material ABS
No. of Compartments 1
Product Style Hinged Lid