Learn More
Electron Microscopy Sciences Laboratory film used to prevent evaporation, contamination and hydration, seals labware quickly and efficiently. Used in EM for drop staining and to pick up thin films when coating grids
SDP
Catalog No. NC2137623
Encompass_Preferred
Laboratory film used to prevent evaporation, conta-mination and hydration. Seals labware quickly and efficiently. Used in EM for drop staining and to pick up thin films when coating grids.
Catalog No. NC2137623 Supplier Electron Microscopy Sciences Supplier No. 70990125
Laboratory film used to prevent evaporation, conta-mination and hydration. Seals labware quickly and efficiently. Used in EM for drop staining and to pick up thin films when coating grids.