Learn More
Electron Microscopy Sciences Parafilm M | 4" x 250' | 1" core
SDP
Catalog No. NC1666852
Encompass_Preferred
Laboratory film used to prevent evaporation, contamination and hydration. Seals labware quickly and efficiently. Used in EM for drop staining and to pick up thin films when coating grids. See recommended dispensers below.
Catalog No. NC1666852 Supplier Electron Microscopy Sciences Supplier No. 70990
Laboratory film used to prevent evaporation, contamination and hydration. Seals labware quickly and efficiently. Used in EM for drop staining and to pick up thin films when coating grids. See recommended dispensers below.