Learn More
Research Products International Corp Parafilm M Laboratory Sealing Film, 4 inch x 250 foot Roll
SDP
Catalog No. 50998944
Encompass
Parafilm M is a semi-transparent, flexible, thermoplastic film that provides a waterproof barrier for covering laboratory vessels. It is gas permeable with low water permeability. Ideal for use in Microbiology and Cell Culture applications as it does not affect oxygen or carbon dioxide permeability. Nontoxic material is resistant to acids, alkalis, alcohols and organic solvents.
Catalog No. 50-998-944 Supplier Research Products International Corp Supplier No. 164999
Parafilm M is a semi-transparent, flexible, thermoplastic film that provides a waterproof barrier for covering laboratory vessels. It is gas permeable with low water permeability. Ideal for use in Microbiology and Cell Culture applications as it does not affect oxygen or carbon dioxide permeability. Nontoxic material is resistant to acids, alkalis, alcohols and organic solvents.