Learn More
Hach Company Parafilm M Laboratory Film
Supplier:  Hach Company 251764
Encompass
Seals labware quickly effectively Film prevents sample evaporation and contamination protects anhydrous materials Can be folded repeatedly and twisted around sharp surfaces without rupturing Stretched lengthwise at 21 C (71 F) Parafilm M increases approximately 200% Becomes more flexible softer stronger as temperature increases Resistant to alcohol mediumconcentration acid and alkalies Width 10 cm (4in ) 38-m (125 ) roll
Catalog No. 50-002-57140