Learn More
Amcor™ Parafilm™ M Laboratory Film
Description
Efficient and cost-effective covering for test tubes, beakers, vials, petri dishes, flasks, and various other instruments and objects with irregular surfaces.
- Protect the integrity of research applications with a tight moisture-barrier that holds a b seal and doesn't release from glassware, despite off-gassing
- Reduce the risk of tampered data with a covering that is odorless, colorless and semi-transparent
- Prevents moisture and volume loss due to material composition and tight seal
- Flexible and self-sealing material allows it to cling around irregular shapes and surfaces
- Stretches more than 200% of original length
- Resistant up to 48 hours against many polar substances such as saline solutions, inorganic acids and alkaline solutions
Permeability
- Oxygen: (ASTM 1927-98) 150 cc/m2 d at 23°C and 50% RH
- Carbon Dioxide: (Modulated IR Method) 1200 cc/m2 d at 23°C and 0% RH
- Water Vapor: (ASTM F1249-01)
- Flat: 1 g/m2 d at 38°C and 90% RH
- Creased: 1 g/m2 d at 38°C and 90% RH
Effects of Common Reagents
- Potassium Permanganate
- 5%: No apparent effect except permanent dark brown coloration in 18 hours
- 0.1%: Same as 5% except slightly less color
- Iodine solution (0.1N)
- No effect except staining brown in 18 hours
- Ethyl Alcohol (95%)
- No apparent effect except some face whitening in 24 hours
- No apparent effect in 24 hours:
- Hydrochloric Acid conc. (12N) dil. (5N)
- Sulphuric Acid conc. (36N) dil. (5N)
- Nitric Acid conc. (16N) dil. (5N)
- Sodium Hydroxide conc. (22%)
- Ammonium Hydroxide conc. (28% NH3)
- Salt (NaCl) solution (20%)
- Isopropyl Alcohol (99%)
- Not recommended for use with chlorinated, non-polar aliphatic and aromatic solvents
- Film becomes soft and sticky at about 130° to 150°F (54° to 66°C)
Order Info
13-374-10, 13-374-12, and 13-374-16 rolls are packaged in cardboard dispensing cartons.
Specifications
Specifications
| Applications | Efficient and cost-effective covering, over wrap to further secure stoppers, lids, and caps, and shelf or tray liner to prevent slipping of bottles or instruments. |
| Length (English) | 125 ft. |
| Color | Natural |
| Description | Amcor PM996 |
| Product Type | Film |
| Width (English) | 4 in. |
| Width (Metric) | 10 cm |
| Length (Metric) | 38 m |
| Material | Thermoplastic |
| Thickness | 5 mil |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.