Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parafilm™ M Wrapping Film
SCI_ED

Catalog No. S37440
Change view
Click to view available options
Length (English):
50 ft.
125 ft.
250 ft.
Width (English):
2 in.
4 in.
20 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Width (English)
S37440 125 ft. 4 in.
S37441 250 ft. 2 in.
S47529 50 ft. 20 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S37440 Supplier Bemis Company Inc Supplier No. PM996/EMD

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Efficient and cost-effective covering for test tubes, beakers, vials, petri dishes, flasks, and various other instruments and objects with irregular surfaces.

  • Protect the integrity of research applications with a tight moisture-barrier that holds a strong seal and doesn’t release from glassware, despite off-gassing
  • Reduce the risk of tampered data with a covering that is odorless, colorless and semi-transparent
  • Prevents moisture and volume loss due to material composition and tight seal
  • Flexible and self-sealing material allows it to cling around irregular shapes and surfaces
  • Stretches more than 200% of original length
  • Resistant up to 48 hours against many polar substances such as saline solutions, inorganic acids and alkaline solutions

Permeability

  • Oxygen: (ASTM 1927-98) 150 cc/m² d at 23°C and 50% RH
  • Carbon Dioxide: (Modulated IR Method) 1200 cc/m² d at 23°C and 0% RH
  • Water Vapor: (ASTM F1249-01)
    • Flat: 1 g/m² d at 38°C and 90% RH
    • Creased: 1 g/m² d at 38°C and 90% RH

Effects of Common Reagents

  • Potassium Permanganate
    • 5%: No apparent effect except permanent dark brown coloration in 18 hours
    • 0.1%: Same as 5% except slightly less color
  • Iodine solution (0.1N)
    • No effect except staining brown in 18 hours
  • Ethyl Alcohol (95%)
    • No apparent effect except some face whitening in 24 hours
  • No apparent effect in 24 hours:
    • Hydrochloric Acid conc. (12N) dil. (5N)
    • Sulphuric Acid conc. (36N) dil. (5N)
    • Nitric Acid conc. (16N) dil. (5nN)
    • Sodium Hydroxide conc. (22%)
    • Ammonium Hydroxide conc. (28% NH<sub>3</sub>)
    • Salt (NaCl) solution (20%)
    • Isopropyl Alcohol (99%)
  • Not recommended for use with chlorinated, non-polar aliphatic and aromatic solvents
  • Film becomes soft and sticky at about 130° to 150°F (54° to 66°C)

Specifications

Applications Efficient and cost-effective covering, over wrap to further secure stoppers, lids, and caps, and shelf or tray liner to prevent slipping of bottles or instruments.
Length (English) 125 ft.
Color Semi-Transparent
Product Type Sealing Film
Width (English) 4 in.
Width (Metric) 10 cm
Material Thermoplastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.