Learn More
Electron Microscopy Sciences Parafilm Parafilm M 2" 250 FT
SDP
Catalog No. 501899425
Encompass_Preferred
Laboratory film used to prevent evaporation, conta-mination and hydration. Seals labware quickly and efficiently. Used in EM for drop staining and to pick up thin films when coating grids.
Catalog No. 50-189-9425 Supplier Electron Microscopy Sciences Supplier No. 70991
Laboratory film used to prevent evaporation, conta-mination and hydration. Seals labware quickly and efficiently. Used in EM for drop staining and to pick up thin films when coating grids.