Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific Parafilm™ Sterile Strips

Catalog No. 01001309
Change view
Click to view available options
Dimensions (L x W):
6 x 4 in.
8 x 2 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
01-001-309 8 x 2 in.
01-001-310 6 x 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-001-309 Supplier Heathrow Scientific Supplier No. 234526ASTL
Add to Cart
Add to Cart

Designed for reliable sealing in laboratory, medical, and industrial environments

  • Ideal for protecting samples and preventing contamination or evaporation perfect for cleanroom environments and strict sanitation protocols
  • These pre-cut strips are sterilized using a Vapor Hydrogen Peroxide process, adding an extra layer of protection to Parafilm natural waterproof and moisture-resistant properties
  • Parafilm™, semi-transparent thermoplastic film designed for reliable sealing in laboratory, medical, and industrial environments
  • Engineered from a unique blend of waxes and polyolefins, Parafilm™ stretches up to 200% original size, conforming tightly to irregular surfaces such as test tubes, beakers, petri dishes, and flasks
  • Packaged in a sealed, lot-traceable pouch featuring a color sterilization indicator ensuring both safety and convenience

Specifications

Length (English) 8 in.
CAS 67-56-1, 63401-08-1
Product Type Parafilm Wrap
Width (English) 2 in.
Width (Metric) 5.08 cm
Length (Metric) 20.3 cm
Material Thermoplastic
Thickness 5 mil
Quantity 50 Pouches, Packs of 10, 1 mL
Dimensions (L x W) 8 x 2 in.
Linear Formula C11H17NO · HCl
MDL Number MFCD00236003
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 215.72
Packaging Ampule
Flash Point 9.7°C (49.5°F)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.