Learn More
Crescent Chemical Co Inc PARAFILM™ 4 X 125
SDP
Supplier:  Crescent Chemical Co Inc SE90310.02
Encompass_Preferred
Parafilm™ 4" x 125' (10 cm x 38 m), roll Roll: width 10 cm (4"), length 38 m (125 ft). Supplied in dispenser box. Size - 6BOX Storage Conditions - +15 °C TO +30 °C Catalog Number - 90310.02
Catalog No. 50-249-233