Learn More
Crescent Chemical Co Inc PARAFILM™ 50 CM X 15 M
SDP
Catalog No. 50249229
Encompass_Preferred
Parafilm™ 50 cm x 15 m, roll Roll: width 50 cm (20"), length 15 m (50 ft). Supplied in sturdy cardboard container. Size - 1ROLL Storage Conditions - +15 °C TO +30 °C Catalog Number - 90300.01
Catalog No. 50-249-229 Supplier Crescent Chemical Co Inc Supplier No. SE90300.01
Parafilm™ 50 cm x 15 m, roll Roll: width 50 cm (20"), length 15 m (50 ft). Supplied in sturdy cardboard container. Size - 1ROLL Storage Conditions - +15 °C TO +30 °C Catalog Number - 90300.01