Learn More
Sigma Aldrich Fine Chemicals Biosciences PARAfILMR M roll size 2 in1EA
Catalog No. 502900469
Encompass
PARAfILM stretches up to 200% and clings even aroundirregularshapesandsurfaces. The sealing film is free of plasticizers and consists primarily of polyolefins and paraffin waxes. PARAfILM M is resistant up to 48 hours against many polar substances eg saline SOLUTIONons inorganic acids and alkaline SOLUTIONons.Protect the integrity of research applications with a tight moisture-barrier that holds a strong seal and doesnt release from glassware despite off-gassingReduce the risk of tampered data with a covering that is odorless colorless and semi-transparentPrevents moisture and volume loss due to material composition and tight sealflexible and self-sealing material allows it to cling around irregular shapes and surfacesStretches more than 200% of original lengthResistant up to 48 hours against many polar substances such as saline SOLUTIONons inorganic acids and alkaline SOLUTIONonsPermeabilityOxygen (ASTM 1927-98) 150 cc/mA²dat 23C and 50%RHCarbon Dioxide (Modulated IR Method) 1200
Catalog No. 50-290-0469 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P75431EA
PARAfILM stretches up to 200% and clings even aroundirregularshapesandsurfaces. The sealing film is free of plasticizers and consists primarily of polyolefins and paraffin waxes. PARAfILM M is resistant up to 48 hours against many polar substances eg saline SOLUTIONons inorganic acids and alkaline SOLUTIONons.Protect the integrity of research applications with a tight moisture-barrier that holds a strong seal and doesnt release from glassware despite off-gassingReduce the risk of tampered data with a covering that is odorless colorless and semi-transparentPrevents moisture and volume loss due to material composition and tight sealflexible and self-sealing material allows it to cling around irregular shapes and surfacesStretches more than 200% of original lengthResistant up to 48 hours against many polar substances such as saline SOLUTIONons inorganic acids and alkaline SOLUTIONonsPermeabilityOxygen (ASTM 1927-98) 150 cc/mA²dat 23C and 50%RHCarbon Dioxide (Modulated IR Method) 1200