Learn More
Sigma Aldrich Fine Chemicals Biosciences ParafilmR M Sealing film f1EA
Catalog No. 502901671
Encompass
Parafilm M Sealing film is colorless moisture-proof film that molds to surface. It is a semi-transparent flexible thermoplastic film with two-way stretch.
Catalog No. 50-290-1671 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS1202651EA
Parafilm M Sealing film is colorless moisture-proof film that molds to surface. It is a semi-transparent flexible thermoplastic film with two-way stretch.