Learn More
Sigma Aldrich Fine Chemicals Biosciences ParafilmR M Sealing film f1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences HS234526B1EA
Encompass
Parafilm(R) M Sealing film is colorless moisture-proof film that molds to surface. It is a semi-transparent flexible thermoplastic film with two-way stretch.
Catalog No. 50-290-0977