Learn More
Andwin Scientific PARAFIN WASTE BAGS 12/CS
SDP
Catalog No. NC0680496
Encompass_Preferred
The Paraffin waste bags are accessories for the Tissue-Tek VIP 6 Vacuum Infiltration Processor, an equipment specially designed for efficient tissue processing. The bags lines the waste container and allows easy removing and disposing after paraffin drainage during a solution exhange. This product is sold in a case of 12.
Catalog No. NC0680496 Supplier Andwin Scientific Supplier No. 6045SAK
The Paraffin waste bags are accessories for the Tissue-Tek VIP 6 Vacuum Infiltration Processor, an equipment specially designed for efficient tissue processing. The bags lines the waste container and allows easy removing and disposing after paraffin drainage during a solution exhange. This product is sold in a case of 12.