Learn More
Electron Microscopy Sciences Paraformaldehyde 1% each Aqueous 10 ML
SDP
Catalog No. 50365725
Encompass_Preferred
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. Filled in 20 ml glass vials (10 ml/vial) allowing for fixation of your specimens without transfer.
Catalog No. 50-365-725 Supplier Electron Microscopy Sciences Supplier No. 1595310
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. Filled in 20 ml glass vials (10 ml/vial) allowing for fixation of your specimens without transfer.