Learn More
Tousimis Researc Corporation Formaldehyde, 10%, MFCD00003274, 10x10mL vials
SDP
Supplier:  Tousimis Researc Corporation 1008C
Encompass
Molecular Formula CH2O, Molecular Weight 30.026, TEM grade, HCHO ultrapure, ready-to-use Packaged under dry-microfiltered nitrogen.
Catalog No. NC9396551