Learn More
Electron Microscopy Sciences Paraformaldehyde 4% 0.1M Phosphate Buffer Saline, pH 7.4 10 ML
SDP
Catalog No. NC0268594
Encompass_Preferred
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. In 0.1M Phosphate Buffer Saline. Filled in 20 ml glass vials (10 ml/vial) allowing for fixation of your specimens without transfer.
pH: 7.4
Catalog No. NC0268594 Supplier Electron Microscopy Sciences Supplier No. 1573585
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. In 0.1M Phosphate Buffer Saline. Filled in 20 ml glass vials (10 ml/vial) allowing for fixation of your specimens without transfer.
pH: 7.4