Learn More
Electron Microscopy Sciences Paraformaldehyde 4%, 0.1M Phosphate Buffer Saline, 500ml
SDP
Supplier:  Electron Microscopy Sciences 1573590
Encompass_Preferred
Paraformaldehyde 4% in 0.1M Phosphate BufferSaline, pH7.4
Catalog No. 50-366-761