Learn More
Sigma Aldrich Fine Chemicals Biosciences Paraformaldehyde, 30525-89-4, MFCD00133991, 500 g
Catalog No. NC1085728
Encompass
Linear Formula: HO(CH2O)nH, Molecular Weight: 30.03 (as monomer), reagent grade, crystalline, MP: 120-170 °C, Synonym: Polyoxymethylene.
Catalog No. NC1085728 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P6148500G
Linear Formula: HO(CH2O)nH, Molecular Weight: 30.03 (as monomer), reagent grade, crystalline, MP: 120-170 °C, Synonym: Polyoxymethylene.