Learn More
Electron Microscopy Sciences Paraformaldehyde is methanol free. It is a more efficient and rapid penetrant fixative that is used in combination with Glutaraldehyde and Acrolein fixatives. CONTAINS NO PRESERVATIVES
SDP
Catalog No. NC2895043
Encompass_Preferred
Paraformaldehyde is methanol free. It is a more efficient and rapid penetrant fixative that is used in combination with Glutaraldehyde and Acrolein fixatives. CONTAINS NO PRESERVATIVES
Catalog No. NC2895043 Supplier Electron Microscopy Sciences Supplier No. 157131G
Paraformaldehyde is methanol free. It is a more efficient and rapid penetrant fixative that is used in combination with Glutaraldehyde and Acrolein fixatives. CONTAINS NO PRESERVATIVES