Learn More
Polysciences, Inc. Paraformaldehyde EM Grade, CAS No.: 30525-89-4, FW: ~/600, MP: 120 deg. to 170 deg.C, Vapor Pressure: 1.45mmHg@25 deg.C 1KG
SDP
Supplier:  Polysciences, Inc. 003801
Encompass_Preferred
Paraformaldehyde EM Grade, CAS No.: 30525-89-4, FW: ~/600, MP: 120 deg. to 170 deg.C, Vapor Pressure: 1.45mmHg@25 deg.C, Depolymerizes in Water to Formaldehyde Solution Yielding Consistent Quality Fixative Solutions, Room Temperature, 1kg
Catalog No. NC9832079