Learn More
Polysciences, Inc. Paraformaldehyde EM Grade, CAS No.: 30525-89-4, FW: ~/600, MP: 120 deg. to 170 deg.C, Vapor Pressure: 1.45mmHg@25 deg.C 250KG
SDP
Catalog No. NC9111107
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
PARAFORMALDEHYDE EM 250G
Catalog No. NC9111107 Supplier Polysciences, Inc. Supplier No. 00380250
This item has a minimum qty of 2 per supplier requirements.
PARAFORMALDEHYDE EM 250G