Learn More
Electron Microscopy Sciences Paraformaldehyde, Granular 1 KG
SDP
Supplier:  Electron Microscopy Sciences 19210
Encompass_Preferred
(Polyoxymethylene)
OH(CH₂O)n H F.W. 600 Average
Paraformaldehyde is the condensation product of methylene glycol, a substance not so far isolated in its pure form; it is a white solid with a formaldehyde content between 78 and 98% and is supplied either as a Prill or Granular. All forms contain more than 95.5% paraformaldehyde content. The Prill and Granular forms are small, free-flowing, dust-free beads, the most desired by EM users.
Catalog No. 50-276-31