Learn More
Sigma Aldrich Fine Chemicals Biosciences Paraformaldehyde reagent g5KG
Catalog No. 502902599
Encompass
Paraformaldehyde is a cross-linking fixative. It is changed to formaldehyde by heating and by adding small amount of sodium hydroxide.
Catalog No. 50-290-2599 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P61485KG
Paraformaldehyde is a cross-linking fixative. It is changed to formaldehyde by heating and by adding small amount of sodium hydroxide.