Learn More
Santa Cruz Biotechnology Paraformaldehyde solution 4% in PBS, 30525-89-4,
Catalog No. NC0238527
Encompass
Molecular Formula C3H6O3X2, Molecular Weight 90.077, Formulation: Contains methanol as a stabilizer. Commonly used to preserve tissues for routine histology in many labs. The solution should be clear, colorless, with no precipitate and the pH 7.0 - 7.6 (25a Deg. C).
Catalog No. NC0238527 Supplier Santa Cruz Biotechnology Supplier No. SC281692
Molecular Formula C3H6O3X2, Molecular Weight 90.077, Formulation: Contains methanol as a stabilizer. Commonly used to preserve tissues for routine histology in many labs. The solution should be clear, colorless, with no precipitate and the pH 7.0 - 7.6 (25a Deg. C).