Learn More
Electron Microscopy Sciences Paraformaldehyde 3% with Glutaraldehyde 0.35% in 0.1M Sodium Cacodylate, pH 7.4 500 ML
SDP
Catalog No. 501901154
Encompass_Preferred
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. Paraformaldehyde 3 % Glutaraldehyde 0.35% in 0.1M Sodium Cacodylate Buffer.
pH: 7.4
Catalog No. 50-190-1154 Supplier Electron Microscopy Sciences Supplier No. 1594960
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. Paraformaldehyde 3 % Glutaraldehyde 0.35% in 0.1M Sodium Cacodylate Buffer.
pH: 7.4