Learn More
Electron Microscopy Sciences Paraformaldehyde 0.5% in 0.1M Phosphate Buffer, pH 7.4 500 ML
SDP
Catalog No. 501901147
Encompass_Preferred
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. In 0.1M Phosphate Buffer.
pH: 7.4
Catalog No. 50-190-1147 Supplier Electron Microscopy Sciences Supplier No. 1595115
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. In 0.1M Phosphate Buffer.
pH: 7.4