Learn More
Electron Microscopy Sciences Paraformaldehyde is methanol free. It is a more efficient and rapid penetrant fixative that is used in combination with Glutaraldehyde and Acrolein fixatives. CONTAINS NO PRESERVATIVES
SDP
Supplier:  Electron Microscopy Sciences 15710250ML
Encompass_Preferred
Paraformaldehyde is methanol free. It is a more efficient and rapid penetrant fixative that is used in combination with Glutaraldehyde and Acrolein fixatives. CONTAINS NO PRESERVATIVES
Catalog No. NC2958033