Learn More
Electron Microscopy Sciences Para/Gard™ Paraffin Repellent 4 OZ
SDP
Supplier:  Electron Microscopy Sciences 6414006
Encompass_Preferred
A paraffin repellent for countertops as well as equipment. Just spray and wipe away. No more paraffin build up around your work area. ParaGard is a pleasant smelling, non-toxic spray which is safe for use on metal, wood, and plastic surfaces.
Catalog No. 50-276-29