Learn More
Cayman Chemical ParaherquamIde E 2.5mg
SDP
Catalog No. 502936229
Encompass_Preferred
A fungal metabolite with anthelmintic and insecticidal activities; lethal to C. elegans (LD50 = 6 μg/ml); lethal to O. fasciatus (LD50 = 0.089 μg/nymph); oral administration reduces T. colubriformis fecal egg count in gerbils from 0.5-4 mg/kg
Catalog No. 50-293-6229 Supplier Cayman Chemical Supplier No. 266802.5 MG
A fungal metabolite with anthelmintic and insecticidal activities; lethal to C. elegans (LD50 = 6 μg/ml); lethal to O. fasciatus (LD50 = 0.089 μg/nymph); oral administration reduces T. colubriformis fecal egg count in gerbils from 0.5-4 mg/kg