Learn More
Electron Microscopy Sciences Veco Parallel Bar with Divider R300D, Nickel
SDP
Catalog No. 5027654
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Nickel
Pitch: 85 µm
Hole: 45 µm
Bar: 38 µm
Catalog No. 50-276-54 Supplier Electron Microscopy Sciences Supplier No. R300DNI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Nickel
Pitch: 85 µm
Hole: 45 µm
Bar: 38 µm