Learn More
Electron Microscopy Sciences Gilder Parallel Bar Grid G400P, Copper
SDP
Catalog No. 5027673
Encompass_Preferred
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.
Material: Copper
Pitch: 62 µm
Hole: 22 µm
Bar: 40 µm
Catalog No. 50-276-73 Supplier Electron Microscopy Sciences Supplier No. G400PCU
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.
Material: Copper
Pitch: 62 µm
Hole: 22 µm
Bar: 40 µm