Learn More
Electron Microscopy Sciences Gilder Parallel Bar Grid G100P, Nickel
SDP
Catalog No. 5027664
Encompass_Preferred
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.
Material: Nickel
Pitch: 250 µm
Hole: 185 µm
Bar: 65 µm
Catalog No. 50-276-64 Supplier Electron Microscopy Sciences Supplier No. G100PNI
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.
Material: Nickel
Pitch: 250 µm
Hole: 185 µm
Bar: 65 µm