Learn More
Electron Microscopy Sciences Veco Parallel Bar Grid R100, Nickel
SDP
Catalog No. 5027641
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Nickel
Pitch: 250 µm
Hole: 200 µm
Bar: 50 µm
Catalog No. 50-276-41 Supplier Electron Microscopy Sciences Supplier No. R100NI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Nickel
Pitch: 250 µm
Hole: 200 µm
Bar: 50 µm