Learn More
Electron Microscopy Sciences Veco Parallel Bar w/ Divider Handle R100D, Nickel
SDP
Catalog No. 5027649
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Material: Nickel
Pitch: 250 µm
Hole: 200 µm
Bar: 50 µm
Catalog No. 50-276-49 Supplier Electron Microscopy Sciences Supplier No. HDR100DNI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Material: Nickel
Pitch: 250 µm
Hole: 200 µm
Bar: 50 µm