Learn More
Electron Microscopy Sciences Veco Parallel Bar w/ Divider Handle R200D, Copper
SDP
Catalog No. 5027647
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Material: Copper
Pitch: 125 µm
Hole: 85 µm
Bar: 40 µm
Catalog No. 50-276-47 Supplier Electron Microscopy Sciences Supplier No. HDR200DCU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Material: Copper
Pitch: 125 µm
Hole: 85 µm
Bar: 40 µm