Learn More
Electron Microscopy Sciences Cold Embedding Parallel Mold, 25 mm
SDP
Catalog No. 501929688
Encompass_Preferred
Mounting cup with parallel sides that help avoid tilting of the mount when clamped into a specimen holder.
Diameter: 25 mm
Catalog No. 50-192-9688 Supplier Electron Microscopy Sciences Supplier No. 5101270
Mounting cup with parallel sides that help avoid tilting of the mount when clamped into a specimen holder.
Diameter: 25 mm